missing translation for 'onlineSavingsMsg'
Learn More

l-Methionine, Sigma-Aldrich™

Product Code. 15820211
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15820211 100 g 100g
15810211 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15820211 Supplier Sigma-Aldrich Supplier No. 8160100100
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-68-3

Specifications

Melting Point 280°C to 282°C
MDL Number MFCD00063097
Synonym L-Methionine, Met, L-Methionine
Density 1.34 g/cm3
Molecular Weight (g/mol) 149.21
Quantity 100 g
Grade Research
Chemical Name or Material L-Methionine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.