missing translation for 'onlineSavingsMsg'
Learn More

l-(-)-Malic Acid, Sigma-Aldrich™

Product Code. 15853991
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15853991 50 g 50g
15843991 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15853991 Supplier Sigma Aldrich Supplier No. 8003840050
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For resolution of racemates for synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-67-6

Specifications

Melting Point 98°C to 103°C
Density 1.60 g/cm3 at 20°C
Quantity 50 g
Molecular Formula HOOCCH(OH)CH2COOH
MDL Number MFCD00064213
Synonym L-(-)-Malic acid, (S)-(-)-Malic acid, L-(-)-Malic acid
Molecular Weight (g/mol) 134.09
Grade Ungraded
Chemical Name or Material L-(-)-Malic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.