missing translation for 'onlineSavingsMsg'
Learn More

L-Lysine [Free Base], MP Biomedicals

Product Code. 11471901 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11471901 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11471901 Supplier MP Biomedicals Supplier No. 0219469680
  • ONLINE EXCLUSIVE PRICE
    882.00 EUR
    840.00 EUR / 100g
    Save 42.00 EUR
    4% Off
 Restricted Item
Estimated Shipment: 09-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 39665-12-8
Molecular Formula C6H16N2O3
MDL Number MFCD00151035
Synonym L-2, 6-Diaminohexanoic acid
InChI Key HZRUTVAFDWTKGD-UHFFFAOYNA-N
SMILES O.NCCCCC(N)C(O)=O
IUPAC Name 2,6-diaminohexanoic acid hydrate
Molecular Weight (g/mol) 164.21
Quantity 100 g
Formula Weight 146.2
Grade Cell Culture Reagent
Physical Form Crystalline
Chemical Name or Material L-Lysine [Free Base]
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.