missing translation for 'onlineSavingsMsg'
Learn More

L-Lysine, 99.6%, MP Biomedicals™

Product Code. 11451681 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11451681 25 g 25g
11441681 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11451681 Supplier MP Biomedicals Supplier No. 0219022425
  • ONLINE EXCLUSIVE PRICE
    148.00 EUR
    140.70 EUR / 25g
    Save 7.30 EUR
    4% Off
 Restricted Item
Estimated Shipment: 09-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 39665-12-8 | C6H16N2O3 | 164.21 g/mol

Chemical Identifiers

CAS 39665-12-8
Molecular Formula C6H16N2O3
Molecular Weight (g/mol) 164.21
MDL Number MFCD00151035
InChI Key HZRUTVAFDWTKGD-UHFFFAOYNA-N
Synonym l-lysine monohydrate, l-lysine hydrate, lysine monohydrate, l +-lysine monohydrate, l-lysine, hydrate, h-lys-oh h2o, lysine, hydrate, lysine hydrate, l-lysine, monohydrate, l-lysine, hydrate 1:1
PubChem CID 16211825
IUPAC Name 2,6-diaminohexanoic acid hydrate
SMILES O.NCCCCC(N)C(O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 164.2
Physical Form Crystalline Powder
Chemical Name or Material L-lysine, Free Base, Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.