Learn More
l-(+)-Lithium Lactate, Sigma-Aldrich™
Product Code. 15891651
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15891651 100 g 100g
15801661 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15891651 Supplier Sigma Aldrich Supplier No. 8220840100
 Restricted Item
Estimated Shipment: 20-11-2026
For synthesis
for synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 27848-80-2
Specifications
Molecular Formula CH3CH(OH)COOLi
MDL Number MFCD00065512
Quantity 100 g
Synonym L-(+)-Lithium lactate, Lactic acid lithium salt, L-(+)-Lithium lactate
Molecular Weight (g/mol) 96.01
Grade Research