Learn More
L-Idaric Acid Disodium Salt, TRC
Product Code. 30397039
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30397039 25 mg 25mg
30398770 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30397039 Supplier Toronto Research Chemicals Supplier No. TRCI16493025MG
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material L-Idaric Acid Disodium Salt
Molecular Formula C6 H8 Na2 O8
Quantity 25 mg
InChI Formula InChI=1S/C6H10O8.2Na/c7-1(3(9)5(11)12)2(8)4(10)6(13)14;;/h1-4,7-10H,(H,11,12)(H,13,14);;/q;2*+1/p-2/t1-,2-,3+,4+;;/m0../s1
SMILES O[C@@H]([C@H](O)[C@@H](O)C(=O)O[Na])[C@@H](O)C(=O)O[Na]
IUPAC Name [(2R,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxo-6-sodiooxyhexanoyl]oxysodium
Molecular Weight (g/mol) 254.102
Formula Weight 254.001