missing translation for 'onlineSavingsMsg'
Learn More

L-Hyocyamine, free base, 100%, MP Biomedicals™

Product Code. 11457971 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 11457971

Brand: MP Biomedicals 0215131601

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 101-31-5
Color White
Assay Percent Range 1
Molecular Formula C17H23NO3
MDL Number MFCD00067306,MFCD00022622,MFCD00022622
Quantity 1 g
Synonym s-atropinium, duboisine, l-tropine tropate, s-atropinium cation, s-atropinium 1+, 3-endo-8-methyl-8-azoniabicyclo 3.2.1 oct-3-yl 2s-3-hydroxy-2-phenylpropanoate, 1r,5s-8-methyl-8-azoniabicyclo 3.2.1 octan-3-yl 2s-3-hydroxy-2-phenylpropanoate, 1r,5s-8-methyl-8-azoniabicyclo 3.2.1 octan-3-yl 2s-3-oxidanyl-2-phenyl-propanoate, 2s-3-hydroxy-2-phenylpropanoic acid 1r,5s-8-methyl-8-azoniabicyclo 3.2.1 octan-3-yl ester
InChI Key RKUNBYITZUJHSG-VFSICIBPSA-N
SMILES CN1[C@H]2CC[C@@H]1CC(C2)OC(=O)[C@H](CO)C1=CC=CC=C1
IUPAC Name (1R,5S)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl (2S)-3-hydroxy-2-phenylpropanoate
Molecular Weight (g/mol) 289.38
PubChem CID 638022
Formula Weight 289.37
Physical Form Crystalline Powder
Chemical Name or Material L-Hyocyamine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.