Learn More
L-Dehydroascorbic acid, 96%
Product Code. 15730849 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 15730849
Brand: Thermo Scientific Acros 250930050
Request Bulk or Custom Format
CAS: 490-83-5 | C6H6O6 | 174.11 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 490-83-5
Molecular Formula C6H6O6
Molecular Weight (g/mol) 174.11
InChI Key SBJKKFFYIZUCET-UHFFFAOYNA-N
IUPAC Name 5-(1,2-dihydroxyethyl)oxolane-2,3,4-trione
SMILES OCC(O)C1OC(=O)C(=O)C1=O
Specifications
Melting Point 228.0°C
Assay Percent Range 95.0 % min. (ELSD) (HPLC)
Quantity 5 g
Formula Weight 174.11
Chemical Name or Material L-Dehydroascorbic acid
compliance-icons
Product Identifier
  • L-Dehydroascorbic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : L-Dehydroascorbic acid