CAS RN 490-83-5
CAS RN 490-83-5
IUPAC Name:
5-(1,2-dihydroxyethyl)oxolane-2,3,4-trione
Synonyms:
dehydroascorbic acid 1-dehydroascorbate L-ascorbic acid, dehydro- acid, dehydroascorbic oxidized ascorbate dehydroascorbates 1-dehydroascorbic acid
Molecular Weight (g/mol):
174.11
Molecular Formula:
C6H6O6
InChi Key:
SBJKKFFYIZUCET-UHFFFAOYNA-N
SMILES:
OCC(O)C1OC(=O)C(=O)C1=O
1
–
2
of
2
results
1
L-Dehydroascorbic acid, 96%
CAS: 490-83-5 Molecular Formula: C6H6O6 Molecular Weight (g/mol): 174.11 InChI Key: SBJKKFFYIZUCET-UHFFFAOYNA-N IUPAC Name: 5-(1,2-dihydroxyethyl)oxolane-2,3,4-trione SMILES: OCC(O)C1OC(=O)C(=O)C1=O
2
Dehydroascorbic acid, MedChemExpress
MedChemExpress Dehydroascorbic acid, a blood-brain barrier transportable form of vitamin C, mediates potent cerebroprotection in experimental stroke.