Learn More
l-Carnitine, Sigma-Aldrich™
Specifications
Specifications
| CAS | 541-15-1 |
| Melting Point | 190°C to 195°C (Decomposes) |
| Density | 0.64 g/cm3 |
| MDL Number | MFCD00038747 |
| Quantity | 25 g |
| Synonym | L-Carnitine, Vitamin B, L-Carnitine |
| Molecular Weight (g/mol) | 161.2 |
| Grade | Research |