Learn More
L-carnitine hydrochloride, 99%, MP Biomedicals™
Product Code. 11490862 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490862 25 g 25g
11470862 1 g 1g
11480862 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11490862 Supplier MP Biomedicals Supplier No. 0219503125
Chemical Identifiers
CAS 6645-46-1
Molecular Formula C7H16ClNO3
Molecular Weight (g/mol) 197.66
MDL Number MFCD00066100
InChI Key JXXCENBLGFBQJM-FYZOBXCZSA-N
Synonym r-3-carboxy-2-hydroxy-n,n,n-trimethylpropan-1-aminium chloride, l-carnitine hcl, unii-j3y5e6iks3, l-carnitine hydrochloride, levocarnitine chloride, j3y5e6iks3, r-carnitine hydrochloride, carnitine l-form hydrochloride, +--carnitine chloride, l-3-carboxy-2-hydroxypropyl trimethylammonium chloride
PubChem CID 656657
IUPAC Name [(2R)-3-carboxy-2-hydroxypropyl]trimethylazanium chloride
SMILES [Cl-].C[N+](C)(C)C[C@H](O)CC(O)=O
Specifications
Color White
Quantity 25 g
Formula Weight 197.7
Percent Purity 0.99
Physical Form Crystalline Powder
Chemical Name or Material L-carnitine hydrochloride