missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartyl-L-Phenylalanine methyl ester, 99.8%, MP Biomedicals™

Product Code. 11454462 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11454462 5 g 5g
11444462 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11454462 Supplier MP Biomedicals Supplier No. 0219000705
  • ONLINE EXCLUSIVE PRICE
    253.00 EUR
    240.45 EUR / 5g
    Save 12.55 EUR
    4% Off
 Restricted Item
Estimated Shipment: 22-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used as a synthetic sweeter, sugar substitute

Chemical Identifiers

CAS 22839-47-0
Molecular Formula C14H18N2O5
Molecular Weight (g/mol) 294.31
MDL Number MFCD00002724
InChI Key IAOZJIPTCAWIRG-QWRGUYRKSA-N
Synonym aspartame, nutrasweet, asp-phe-ome, asp-phe methyl ester, aspartam, aspartamum, aspartamo, l-aspartyl-l-phenylalanine methyl ester, aspartylphenylalanine methyl ester, dipeptide sweetener
PubChem CID 134601
ChEBI CHEBI:2877
IUPAC Name (3S)-3-amino-3-{[(2S)-1-methoxy-1-oxo-3-phenylpropan-2-yl]carbamoyl}propanoic acid
SMILES COC(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@@H](N)CC(O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 294.31
Percent Purity ≥98%
Grade Research
Physical Form Powder
Chemical Name or Material L-Aspartyl-L-Phenylalanine methyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.