Learn More
L-Aspartyl-L-Phenylalanine methyl ester, 99.8%, MP Biomedicals™
Product Code. 11444462 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11444462
Brand: MP Biomedicals 0219000701
Used as a synthetic sweeter, sugar substitute
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 22839-47-0
Molecular Formula C14H18N2O5
Molecular Weight (g/mol) 294.31
MDL Number MFCD00002724
InChI Key IAOZJIPTCAWIRG-QWRGUYRKSA-N
Synonym aspartame, nutrasweet, asp-phe-ome, asp-phe methyl ester, aspartam, aspartamum, aspartamo, l-aspartyl-l-phenylalanine methyl ester, aspartylphenylalanine methyl ester, dipeptide sweetener
PubChem CID 134601
ChEBI CHEBI:2877
IUPAC Name (3S)-3-amino-3-{[(2S)-1-methoxy-1-oxo-3-phenylpropan-2-yl]carbamoyl}propanoic acid
SMILES COC(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@@H](N)CC(O)=O
Specifications
Color White
Quantity 1 g
Formula Weight 294.31
Physical Form Powder
Chemical Name or Material L-Aspartyl-L-Phenylalanine methyl ester