Learn More
L-Alanyl-L-Alanyl-L-Alanyl-L-Alanine, TRC
Product Code. 30179460
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179460 50 mg 50mg
30179406 100 mg 100mg
30179620 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30179460 Supplier Toronto Research Chemicals Supplier No. TRCA57648850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 926-79-4
Synonym Alanine quadramer∣L-Ala-L-Ala-L-Ala-L-Ala∣L-Alanine tetramer∣Tetra-L-alanine∣Tetraalanine
Specifications
Format Neat
Molecular Formula C12H22N4O5
MDL Number MFCD00066035
Molecular Weight (g/mol) 302.33
InChI Key ZHRZLXZJVUFLNY-XAMCCFCMSA-N
SMILES C[C@H](N)C(=O)N[C@@H](C)C(=O)N[C@@H](C)C(=O)N[C@@H](C)C(O)=O
IUPAC Name (2S)-2-[(2S)-2-[(2S)-2-[(2S)-2-aminopropanamido]propanamido]propanamido]propanoic acid
Quantity 50 mg
Shipping Condition Room Temperature