Learn More
Thermo Scientific Chemicals L-(+)-2-Phenylglycine, 98+%
Product Code. 11433967
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433967 25 g 25g
11443967 100 g 100g
11408860 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11433967 Supplier Thermo Scientific Chemicals Supplier No. A19360.14
 Restricted Item
In Stock
Add to basket
CAS: 2935-35-5 | C8H9NO2 | 151.165 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2935-35-5
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00064403
InChI Key ZGUNAGUHMKGQNY-ZETCQYMHSA-N
Synonym h-phg-oh, l-phenylglycine, s-2-amino-2-phenylacetic acid, l-+-alpha-phenylglycine, l-2-phenylglycine, l-+-2-phenylglycine, s-phenylglycine, phenylglycine, 2s-2-amino-2-phenylacetic acid, 2s-amino phenyl acetic acid
PubChem CID 99291
ChEBI CHEBI:439819
IUPAC Name (2S)-2-amino-2-phenylacetic acid
SMILES C1=CC=C(C=C1)C(C(=O)O)N
Specifications
Melting Point 288°C (decomposition)
Beilstein 2208675
Merck Index 14,7291
Quantity 25 g
Formula Weight 151.17
Optical Rotation +156° (c=1 in 1N HCl)
Percent Purity ≥98%
Chemical Name or Material L-(+)-2-Phenylglycine