Learn More
Saint Gobain Life Sciences Kynar PVDF Bag
Product Code. 13345211
Change view
Click to view available options
Capacity (Metric):
8 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
13345211 8 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13345211 Supplier Saint Gobain Life Sciences Supplier No. 0141012
Estimated Shipment: 21-01-2027
Offer an economical alternative for analysis of noncorrosive gases. Kynar PVDF Bag features a corrosive-resistant Kynar construction for all your sampling application needs.
  • Flexible
  • Will not contaminate sample
  • For use in gas sampling trains
  • Use for gas or liquid sample containment
  • Durable, heat-sealed seams ensure leak-proof operation
TRUSTED_SUSTAINABILITY
Specifications
Material PVDF
Thickness (English) 0.003 in.
Dimensions (L x W) 305 x 305 mm
Length (English) 12 in.
Width (English) 12 in.
Product Type Kynar Bag
Capacity (Metric) 8 L
Length (Metric) 305 mm
Width (Metric) 305 mm
Thickness (Metric) 0.070 mm