Learn More
DWK Life Sciences Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support and Ground Joint (90mm)
Product Code. 11877513
Change view
Click to view available options
Volume (Metric):
1 L
2 L
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11877513 2 L Each
11867513 1 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11877513 Supplier DWK Life Sciences Supplier No. 9538350090
 Restricted Item
Estimated Shipment: 27-05-2027
Add to basket
Add to basket
Designed for large volume filtration analysis of corrosive liquids or samples with high particulate loads that would tend to clog a 47 mm diameter filter
This unit is also recommended for filtering large volumes of HPLC solvents.
  • The ground glass connection eliminates the possibility of phthalate contamination that can occur when using silicone or neoprene stoppers
      Each apparatus is supplied with 1000mL funnel; 90mm, 40-60 micron porosity, fritted glass support base; anodized aluminum clamp; and filtration flask
    • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Material Glass
Type Microfiltration Assembly
Volume (Metric) 2 L