Learn More
DWK Life Sciences KIMBLE™ KONTES™ PTFE Tubing
Product Code. 15370158
Change view
Click to view available options
Diameter (English) Inner:
0.031 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Inner Unit Size
15370158 0.031 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15370158 Supplier DWK Life Sciences Supplier No. 4208230116
  • ONLINE EXCLUSIVE PRICE
    36.85 EUR
    29.06 EUR / Pack of 1
    Save 7.79 EUR
    21% Off
View all promotions
 Restricted Item
Estimated Shipment: 05-02-2027
KIMBLE™ KONTES™ PTFE Tubing is chemically inert and suitable for low pressure applications. Precision outer diameter tolerances ensure reliable connections with flangeless nuts and ferrules, and the tubing is designed for easy connection to Flex-Columns™.
Chemically inert; suitable for low pressure applications
  • Chemically inert PTFE construction suitable for low pressure applications
  • Outer diameter tolerances of the 0.063 in. O.D. tubing held at ±0.001 in., assuring a good connection with flangeless nuts and ferrules
  • Designed for easy connection to Flex-Columns™
  • Available in 10 ft. lengths
Specifications
Type Tubing
Diameter (English) Outer 0.063 in.
Diameter (English) Inner 0.031 in.
For Use With (Application) Suitable for low pressure applications
Overall Length (English) 10 ft.
Material PTFE