Learn More
Pacific Handy Cutter Kleen™ Antimicrobial Enclosed Blade Safety Cutter
Description
- Unlike most utility knives and cutters with exposed blades, the blade in the Kleen™ EBC1X is set back into the head of the cutter, providing complete protection to the user.
- For added safety, this convenient tool features a metal bladeless tape splitter which allows users to open boxes without even utilizing a blade, which also helps prevent damaging the contents of boxes.
- The versatile EBC1 effortlessly cuts corrugated and cardboard boxes, stretch/shrink wrap, plastic strapping, box tape, and much more.
- It also features a lanyard hole for tethering to a PHC lanyard for better retention.
Specifications
Specifications
| Product Type | Kleen Knife |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Color | Green |
Product Title
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?