Learn More
SCHOTT AG Lighting and Imaging™ KL 300 LED Cold Light Source
Product Code. 12599816 Shop All Schott Products
Change view
Click to view available options
Range (Lux):
80 Lm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range (Lux) Unit Size
12599816 80 Lm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 12599816 Supplier SCHOTT AG Lighting and Imaging™ Supplier No. 120300
Power Your Renewable Energy Research
Use promo code 28029
View all promotions
Estimated Shipment: 22-12-2026
Regulatory: This product is under eco-contribution tax.
Innovative fiber optic LED Light Source uses cold light to illuminate all types of objects in life sciences and industrial applications. The brightness equivalent to a 30-watt halogen lamp is ideal as a standard illumination system for the requirements of routine inspections and education.
  • 80 Lm lightflux.
  • No fan, silent operation.
  • Compact ergonomical design.
  • Mountable direct to the stand or pillar.
  • Adaptors for all stands plus stand-alone version available.
  • Mounting bracket need to be ordered separately.
  • Wide range power supply included with international clip plug system.
  • Fiber optic light guides or ringlights need to be ordered separately.
Specifications
Type Cold Light Source
Certifications/Compliance CE Certified, B EMC-Class
Dimensions (L x W x H) 107 x 114 x 61 mm
For Use With (Application) Stereo Microscopy Illumination
For Use With (Equipment) SCHOTT Fiber Optic Flexible Lightguides, Goosenecks or Ringlights with active lightguide diameter of max. 6 mm
Frequency 50/60 Hz
Height (English) 2.4 in.
Height (Metric) 61 mm
Includes Wide range power supply with international clip plug system
Lamp Life 50,000 Hours
Length (English) 4.2 in.
Length (Metric) 107 mm
Lighting Type 1 High Power LED white
Range (Lux) 80 Lm
Voltage 100/240 V
Width (English) 4.5 in.
Width (Metric) 114 mm