Learn More
Lenz Laborglasintrumente Kjeldahl Flasks
Product Code. 10604392
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
750 mL
1 L
Unit Size:
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10604392 1 L Pack of 10
10315361 250 mL Pack of 10
10264621 100 mL Pack of 10
10420323 750 mL Pack of 10
10570112 50 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10604392 Supplier Lenz Laborglasintrumente Supplier No. 03052970
  • ONLINE EXCLUSIVE PRICE
    1025.00 EUR
    798.60 EUR / Pack of 10
    Save 226.40 EUR
    22% Off
Estimated Shipment: 28-12-2026
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • For the determination of nitrogen in organic compounds
  • With center neck
  • With conical joint and inscription label
  • Material: DURAN™
Specifications
Capacity (Metric) 1 L
Material Borosilicate Glass (3.3)
Standard Taper Joint NS 29/32
Diameter (Metric) Outer 126 mm
Quantity 10/Pk.