Description
Optimize performance using Thermo Scientific™ KingFisher™ Systems Consumables, specifically designed for use with the KingFisher System and ideal for magnetic particle processing because of their low binding affinity for biomolecules. The specially-designed, disposable tip combs and microplates, which are made of polypropylene, enable excellent recovery of magnetic beads.
- Specifically designed for use with the KingFisher System
- Ideal for magnetic particle processing because of their low binding affinity for biomolecules
- The specially-designed, disposable tip combs and microplates are made of polypropylene
- This enables excellent recovery of magnetic beads
Specifications
Specifications
| Capacity | 200 μL |
| For Use With (Application) | Magnetic particle processing |
| For Use With (Equipment) | KingFisher System |
| Includes | KingFisher plate 200 μL |
| Quantity | 50/Cs. |