Learn More
MedChemExpress Kinetensin
Product Code. 16489528
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16489528 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16489528 Supplier MedChemExpress Supplier No. HYP1255
 Restricted Item
Estimated Shipment: 29-09-2026
MedChemExpress Kinetensin is a neurotensin-like peptide isolated from pepsin-treated human plasma.
Specifications
Purity 99.21%
CAS 103131-69-7
Content And Storage Protect from light, stored under nitrogen∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light, stored under nitrogen)
For Use With (Application) Neuroscience-Neuromodulation
Molecular Weight (g/mol) 1172.38 g/mol
Product Type Natural Products
Quantity 10 mg
Shipping Condition Room temperature
Solubility DMSO : 100 mg/mL (85.30 mM∣Need ultrasonic)
Synonym Kinetensin (human)