Learn More
DWK Life Sciences Kimble™ Kontes™ Closed System Sterile Tissue Grinder (Molded Tip)
Product Code. 12643225
Change view
Click to view available options
Capacity (Metric):
15 mL
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12643225 15 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12643225 Supplier DWK Life Sciences Supplier No. 7496000015
  • ONLINE EXCLUSIVE PRICE
    181.50 EUR
    135.99 EUR / Pack of 10
    Save 45.51 EUR
    25% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Includes
Fully assembled, 30% glass-filled pestles, sample tubes with caps, and adhesive labels.
Minimizes the risk of personal contact with samples
  • Tissues are ground within a sealed container
  • Narrow pestles eliminate the potential of sample overflow
  • Molded-in abrasive surface on the pestle tip leaves no sediment to obstruct sample examination
  • Gamma sterilized in easy-open HDPE packs
  • CS2 tissue grinders have a vitrified tip for more aggressive grinding of difficult samples
  • Polypropylene pestles and conical sample tubes with solid HDPE caps
Specifications
Capacity (English) 0.5 oz.
Capacity (Metric) 15 mL
Includes Fully assembled, 30% Glass filled Polypropylene pestle, Polypropylene conical sample tube with solid HDPE cap and an adhesive label