Learn More
DWK Life Sciences Kimble™ KIMAX™ Iodine Determination Flasks with PTFE Stopper
For analysis of dry shellac, varnishes, oils, fatty acids
703.95 EUR - 1011.24 EUR
Specifications
Closure Material | PTFE |
---|---|
Material | Borosilicate Glass |
Writing Surface | Matte Finish Ceramic Enamel Marking |
Closure Size | 22 |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11823133
|
DWK Life Sciences
27200500 |
500 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11813133
|
DWK Life Sciences
27200250 |
250 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11803133
|
DWK Life Sciences
27200125 |
125 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
For analysis of dry shellac, varnishes, oils, fatty acids
250mL size
- Analyze dry shellac and shellac varnishes according to ASTM™ D 29
- Determine total iodine value of oils, fatty acids, and bodied oils according to ASTM D 1541
- Test drying oils for use in paints and varnishes according to ASTM D 555
Compliance
Designed from ASTM E 1404, Type II, Class 3 requirements
Specifications
PTFE | |
Borosilicate Glass | |
Matte Finish Ceramic Enamel Marking | |
22 |