Learn More
DWK Life Sciences Kimble™ KIMAX™ Iodine Determination Flasks with PTFE Stopper
For analysis of dry shellac, varnishes, oils, fatty acids
Brand: DWK Life Sciences 27200250
Additional Details : Weight : 0.80000kg
Description
For analysis of dry shellac, varnishes, oils, fatty acids
- PTFE standard taper stopper is included
- A durable matte-finish ceramic enamel marking spot takes writing and erases easily
- Designed from ASTM Specification E1404, Type II, Class 3 requirements.
Specifications
PTFE | |
250 mL | |
6.29 in. | |
Matte Finish Ceramic Enamel Marking | |
22 |
3.2 in. | |
Borosilicate Glass | |
16 cm | |
82 mm |