Learn More
DWK Life Sciences Kimble™ KIMAX™ Iodine Determination Flasks with PTFE Stopper
For analysis of dry shellac, varnishes, oils, fatty acids
Marque:  DWK Life Sciences 27200500
Informations supplémentaires : Weight : 0.80000kg
 Afficher plus de versions de ce produit
Code produit. 11823133
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
For analysis of dry shellac, varnishes, oils, fatty acids
  • PTFE standard taper stopper is included
  • A durable matte-finish ceramic enamel marking spot takes writing and erases easily
  • Designed from ASTM Specification E1404, Type II, Class 3 requirements.
Spécification
Spécification
PTFE
500 mL
7.67 in.
Matte Finish Ceramic Enamel Marking
22
4.01 in.
Borosilicate Glass
19.5 cm
102 mm
Suggestions de produits
Suggestions de produits
Vidéos
FDS
Documents
Documents
Certificats