Learn More
DWK Life Sciences KIMBLE™ EPA Water Analysis Vial, Without Closure
Product Code. 13111643 Shop All DWK Life Sciences Products
Change view
Click to view available options
Volume (Metric):
20 mL
30 mL
40 mL
60 mL
Color:
Clear
No. per Pack:
144
Unit Size:
Pack of 144
Pack of 432
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Color No. per Pack Unit Size
13111643 60 mL Clear 144 Pack of 144
12703077 20 mL Clear 144 Pack of 432
12713077 30 mL Clear 144 Pack of 432
13080013 40 mL Clear 144 Pack of 432
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 13111643 Supplier DWK Life Sciences Supplier No. 60958A16
  • ONLINE EXCLUSIVE PRICE
    630.00 EUR
    589.53 EUR / Pack of 144
    Save 40.47 EUR
    6% Off
 Restricted Item
Estimated Shipment: 22-09-2027
The KIMBLE™ EPA Water Analysis Vial, Without Closure is designed for collection and storage of water samples. Available in clear and amber borosilicate glass, these screw thread vials comply with EPA 40 CFR 136 and 40 CFR 141 guidelines for the analysis of pollutants.
  • Clear EPA vials are manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
  • Amber EPA vials are converted from Type I, 51 expansion borosilicate glass tubing
  • Comply with EPA 40 CFR 136 guidelines for establishing test procedures for the analysis of pollutants and EPA 40 CFR 141 National Interim Primary Drinking Water Regulations
  • Vials are packaged lehr-clean in a plastic shrink wrap module and placed into a corrugated shelf-pack
  • Open-top closures with PTFE-faced silicone septa or solid-top PTFE-faced white rubber-lined closures are available separately
  • 24-400 GPI screw thread finish
Compliance
Vials comply with guidelines for establishing test procedures for analyzing pollutants in water (Ref. U.S. EPA Methods 601, 602, 603, and 604).
Specifications
Diameter (English) Outer 1.18 in.
Color Clear
Diameter (Metric) Outer 30 mm
Material Glass (Type 1)
Sterility Non-sterile
Volume (Metric) 60 mL
For Use With (Application) For water analysis
Height (English) 4.84 in.
No. per Pack 144
Volume (English) 2.02 oz.
Closure Type Screw Cap
Quantity 432/Cs.
Marking Spot No
Autoclavable Autoclavable
Certifications/Compliance EPA 601, 602, 603, 604
Height (Metric) 123 mm
Preservative None
Product Type Vial