Learn More
DWK Life Sciences Kimble™ Closures for Sample Vials
Product Code. 17781078
Click to view available options
Closure Size:
13-425
15-425
18-400
20-400
24-400
Unit Size:
Pack of 144
Product Code. 17781078
Brand: DWK Life Sciences 7380218400
Black phenolic closure with PTFE-lined cap
  • Works with media bottles and screw-thread vials
  • Special black phenolic material (wood filled) very resistant to the effects of temperature and steam encountered in autoclaving
  • Cemented-in PTFE-faced rubber liner is provided
TRUSTED_SUSTAINABILITY
Specifications
Closure Size 18-400
Liner Material PTFE, Rubber
Product Type Screw Cap
Material Phenolic
Color Black