Learn More
DWK Life Sciences Kimble™ Kontes™ Sterile Disposable Pasteur Pipet
Description
SteriPettes are sterile, disposable Pasteur pipets.
- Gamma-irradiated to assure sterility
- Convenient 250 piece multi-pack configuration
- 10 peel packs of 25 each
- Available in both 5-3/4 in. and 9 in. lengths
- Constricted top for cotton plugging
Specifications
Specifications
| Certifications/Compliance | ASTM E732-80, USP Type III |
| Diameter (English) Inner | Tip Opening: 0.043 in. |
| Diameter (Metric) Inner | Tip Opening: 0.11 cm |
| Material | Soda Lime Glass |
| Sterility | Sterile |
| For Use With (Application) | For IVF procedures |
| Length (English) | 9 in. |
| Volume (English) | 0.067 oz. |
| Volume (Metric) | 2 mL |
| Length (Metric) | 22.86 cm |
| Show More |