Learn More
DWK Life Sciences Kimble™ Kontes™ Right Angle without Tip O-Ring PTFE Plug HI-VAC Valves
Product Code. 11702235
Change view
Click to view available options
Range:
0 to 12 mm
0 to 4 mm
Bore Size (English):
0 to 0.15 in.
0 to 0.47 in.
Diameter (English) Outer Stem:
0.3 in.
0.62 in.
Shaft O-Ring Size:
010
112
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Bore Size (English) Diameter (English) Outer Stem Shaft O-Ring Size unitSize
11702235 0 to 12 mm 0 to 0.47 in. 0.62 in. 112 Each
11792225 0 to 4 mm 0 to 0.15 in. 0.3 in. 010 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Polyacetal knob is not recommended for use over 110°C.
Product Code. 11702235 Supplier DWK Life Sciences Supplier No. 8266100012
 Restricted Item
Estimated Shipment: 19-04-2027
Add to basket
Add to basket
Polyacetal knob is not recommended for use over 110°C.
For vacuum applications up to 5 x 10-7 torr
  • Comes with precision external thread for fine control
  • Non-rotating shaft with primary seal made with FKM o-ring on tapered seat
  • Full open position at 1-1/2 turns
  • Used as bleed at up to half turn
  • PTFE shaft can be used continuously at system temperatures of 230°C or intermittently at 275°C
TRUSTED_SUSTAINABILITY
Specifications
Bore Size (English) 0 to 0.47 in.
Diameter (English) Outer Stem 0.62 in.
Bore Size (Metric) 0 to 1.2 cm
Diameter (Metric) Outer Stem 1.6 cm
Includes PTFE plug
Material Borosilicate Glass
Shaft O-Ring Size 112
Range 0 to 12 mm
Type Right Angle Valve