Learn More
DWK Life Sciences KIMBLE™ KONTES™ Pestles For Dounce All-Glass Tissue Grinders
Product Code. 13161124 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (English):
0.06 oz.
0.23 oz.
Capacity (Metric):
2 mL
7 mL
Product Type:
Large Clearance Pestle, A
Small Clearance Pestle, B
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (English) Capacity (Metric) Product Type Unit Size
13161124 0.06 oz. 2 mL Large Clearance Pestle, A Each
10389444 0.06 oz. 2 mL Small Clearance Pestle, B Each
10581133 0.23 oz. 7 mL Small Clearance Pestle, B Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 13161124 Supplier DWK Life Sciences Supplier No. 8853010002
  • ONLINE EXCLUSIVE PRICE
    56.50 EUR
    53.33 EUR / Pack of 1
    Save 3.17 EUR
    5% Off
 Restricted Item
Estimated Shipment: 08-06-2027
KIMBLE™ KONTES™ Pestles For Dounce All-Glass Tissue Grinders are all-glass replacement pestles designed for use with Dounce (KONTES™ 885300 series) tissue grinders. Available in Pestle A (large clearance) and Pestle B (small clearance) configurations.
  • Replacement glass pestles for use with Dounce (Kontes 885300 series) tissue grinders
  • Available in Pestle A (large clearance: 0.0030-0.0050in.) and Pestle B (small clearance: 0.0005-0.0025in.) configurations
  • Manufactured from 33 expansion, low extractable borosilicate glass
  • Conforms to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Capacity (English) 0.06 oz.
Capacity (Metric) 2 mL
Material Borosilicate Glass
Product Type Large Clearance Pestle, A
For Use With (Equipment) Dounce tissue grinder