Learn More
DWK Life Sciences Kimble™ Kontes™ Pellet Pestle™
Product Code. 11898502
Change view
Click to view available options
Capacity (Metric):
0.5 mL
1.5 mL
Material:
CTFE/Stainless Steel
Polypropylene
Includes:
100 Microtubes
w/o Microtubes
Dnase Free:
No
RNase, DNase and Pyrogen-free
Unit Size:
Each
Pack of 100
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Material Includes Dnase Free Unit Size
11898502 1.5 mL CTFE/Stainless Steel w/o Microtubes No Each
11888502 0.5 mL CTFE/Stainless Steel w/o Microtubes No Each
11872913 1.5 mL Polypropylene 100 Microtubes No Pack of 100
11858502 0.5 mL Polypropylene w/o Microtubes RNase, DNase and Pyrogen-free Pack of 100
10057312 1.5 mL Polypropylene 100 Microtubes RNase, DNase and Pyrogen-free Pack of 100
10794783 0.5 mL Polypropylene 100 Microtubes RNase, DNase and Pyrogen-free Pack of 100
11815125 1.5 mL Polypropylene w/o Microtubes RNase, DNase and Pyrogen-free Pack of 100
11800104 0.5 mL Polypropylene w/o Microtubes No Pack of 100
11849923 1.5 mL Polypropylene w/o Microtubes No Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 11898502 Supplier DWK Life Sciences Supplier No. 7495150000
  • ONLINE EXCLUSIVE PRICE
    125.40 EUR
    99.92 EUR / Pack of 1
    Save 25.48 EUR
    20% Off
 Restricted Item
Estimated Shipment: 16-04-2027
Use to resuspend protein and DNA pellets or grind soft tissue in microcentrifuge tubes
Disposable Pellet Pestle
  • Choose autoclavable polypropylene or RNase-, DNase-, pyrogen-free material (manufactured under M5.5 [Class 10,000] cleanroom conditions)
  • Available with or without 100 microtubes
  • Microtubes have the same properties and come individually wrapped to ensure purity
  • Length of pestle with tip, 7cm (2.75 in.)
Reusable Pellet Pestle
  • Autoclavable CTFE polymeric head with a stainless-steel shaft
  • Pestle length with tip, 6cm (2.5 in.) for 0.5mL size; 14cm (5.59 in.) for 1.5mL size
Specifications
Autoclavable Autoclavable
Capacity (English) 0.05 oz.
Capacity (Metric) 1.5 mL
Material CTFE/Stainless Steel
Product Type Pellet Pestle
Disposable Reusable
For Use With (Application) Resuspend Protein and DNA pellets or grind soft tissue in microcentrifuge tubes
Length (English) 0.55 in.
Dnase Free No
Includes w/o Microtubes
For Use With (Equipment) 500μL and 1.5mL microcentrifuge tube
Length (Metric) 14mm
Length (Metric) Tube 14mm