Learn More
DWK Life Sciences Kimble™ Kontes™ Microcaps™ TLC Spotting Capillaries
Product Code. 13075813
Change view
Click to view available options
Volume (Metric):
0.5, 1, 2, 5, 10 μL
100 μL
20 μL
5 μL
Unit Size:
Pack of 100
Pack of 500
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
13075813 0.5, 1, 2, 5, 10 μL Pack of 500
13005813 5 μL Pack of 100
13025813 20 μL Pack of 100
13065813 100 μL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 13075813 Supplier DWK Life Sciences Supplier No. 7645200000
  • ONLINE EXCLUSIVE PRICE
    246.75 EUR
    232.51 EUR / Pack of 500
    Save 14.24 EUR
    5% Off
 Restricted Item
Estimated Shipment: 28-01-2027
For spotting preadsorbent TLC plates
Kit includes:
  • Five dispenser vials containing 100 micropipets each of 0.5, 1, 2, 5 and 10μL
  • Bulb assembly
  • Spotting holder
Specifications
Material Borosilicate Glass
Includes Five dispenser vials containing 100 micropipets each of 0.5, 1, 2, 5 and 10μL; Bulb assembly; Spotting holder
Volume (Metric) 0.5, 1, 2, 5, 10 μL
Type TLC Spotting Capillary Kit