Learn More
DWK Life Sciences KIMBLE™ KIMAX™ Erlenmeyer Flasks with Screw Caps
Product Code. 11797979 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Pack of 2
Pack of 4
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11797979 500 mL Pack of 6
11787979 250 mL Pack of 6
11707989 1000 mL Pack of 4
11717989 2000 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11797979 Supplier DWK Life Sciences Supplier No. KC26720500
  • ONLINE EXCLUSIVE PRICE
    351.00 EUR
    330.80 EUR / Pack of 6
    Save 20.20 EUR
    5% Off
 Restricted Item
Estimated Shipment: 26-02-2027
Includes
Blue caps and pour rings
KIMBLE™ KIMAX™ Erlenmeyer Flasks with Screw Caps feature an autoclavable KimCote™ plastic safety coating to reduce hazards of shattered glass. GL 45 threaded closures ensure tight seals. Blue polypropylene caps and clear pour rings included.
Plastic-coated for added safety
  • Autoclavable KimCote™ plastic safety coating reduces hazards of shattered glass and leakage of toxic or corrosive chemicals
  • Large openings with GL 45 threaded closures for easy filling and tight sealing during storage
  • Blue polypropylene caps and clear pour rings included
  • Marking spots and graduation scales in durable white ceramic enamel
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
  • Suitable for mixing and storing culture media and other culturing techniques
Specifications
Closure Color Blue
Closure Material Polypropylene
Description Reverse metric scale graduations
Diameter (English) Outer 4.01 in.
Capacity (Metric) 500 mL
Material Borosilicate Glass
Graduation Range 25 to 500 mL
Height (English) 6.5 in.
Height (Metric) 16.5 cm
Includes Clear pour rings
Product Type Erlenmeyer Flask
Writing Surface White Ceramic Enamel Marking
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Mouth Style Threaded
Sterility Non-sterile
Bottom Shape Conical
Closure Type Screw
Diameter (Metric) Outer 102 mm
Surface Treatment Untreated