Learn More
DWK Life Sciences Kimble™ KIMAX™ Tall-Form Beakers, with spout
Product Code. 31925057
Change view
Click to view available options
Capacity (Metric):
300 mL
Unit Size:
Pack of 12
Pack of 48
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
31925057 300 mL Pack of 48
11740704 300 mL Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 31925057 Supplier DWK Life Sciences Supplier No. 14030300
With spout for easy pouring.
  • Matte-finish marking spot
  • Graduated to indicate approximate volumes
  • Berzelius style with flared top, reinforcing bead
Compliance
Designed from ASTM™ E 960 Type III (with spout) and Type IV (without spout) requirements
TRUSTED_SUSTAINABILITY
Specifications
Capacity (English) 10.14 oz.
Diameter (English) Outer 2.51 in.
Diameter (Metric) Outer 6.4 cm
Capacity (Metric) 300 mL
Material Borosilicate Glass
For Use With (Application) For Performing Titrations
Graduation Range 25 to 250 mL
Height (English) 4.64 in.
Height (Metric) 11.8 cm
Includes Spout
Increments 25 mL
Label Type Matte Finish Marking
Quantity 12 Pk.
Type Beaker
Disposable Reusable
Graduated Yes
Certifications/Compliance ASTM E960, Type III, USP Type I, ASTM E438, Type I, Class A
Scale Dual White Scale
Style Tall Flared Top