Learn More
DWK Life Sciences KIMBLE™ KIMAX™ Borosilicate Glass Aspirator Bottles with Tubulation
Product Code. 11876081
Change view
Click to view available options
Capacity (Metric):
1000 mL
Stopper Number:
6
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Stopper Number Unit Size
11876081 1000 mL 6 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11876081 Supplier DWK Life Sciences Supplier No. 146071000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Benötigt
Tubing
KIMBLE™ KIMAX™ Borosilicate Glass Aspirator Bottles with Tubulation are made from 33 expansion borosilicate glass with a tooled top, narrow mouth for a rubber stopper, and a white enamel marking spot. Available in multiple capacities for reliable liquid storage and dispensing.
Bottom glass hose-connection outlet is fused to bottle
  • Tooled top for a secure, uniform fit
  • Narrow mouth designed to accept a rubber stopper
  • White enamel marking spot for easy labeling
  • Manufactured from 33 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
  • Available in multiple capacities from 250 mL to 20 L
Bestellinformation
8.5 oz. (250mL) and 17 oz. (500mL) sizes accept 0.312 in. I.D. (8mm) tubing. 34 oz. (1L) and larger accept 0.375 in. I.D. (9.5mm) flexible tubing.
Compliance
Meet ASTM™ E 438 Type I, Class A requirements.
Spezifikation
Capacity (Metric) 1000 mL
Color Clear
Diameter (English) 4.33 in.
Diameter (English) Inner Neck 1.06 in.
Diameter (Metric) Inner Neck 27 mm
For Use With (Application) Designed to store and discharge liquids via a bottom hose outlet
Includes Bottom hose outlet, Marking Spot
Material Borosilicate Glass
Sterility Non-sterile
Stopper Number 6
Mouth Narrow
Height (Metric) 200 mm
Height (English) 7.87 in.
Diameter (Metric) 110 mm
Diameter (English) Inner Tubing 0.312 in.
Diameter (Metric) Inner Tubing 7.92 mm
Autoclavable Not Autoclavable
Capacity (English) 0.25 gal.
Product Type Reservoir Bottle
Mehr anzeigen Weniger anzeigen