Learn More
DWK Life Sciences Kimble™ Kim-Kap™ Closures
Product Code. 10399924
Change view
Click to view available options
Color:
Blue
Green
Natural
Red
Yellow
For Use With (Equipment):
13 mm Plain-end Culture Tubes
16 mm Plain-end Culture Tubes
18 mm Plain-end Culture Tubes
20 mm Plain-end Culture Tubes
25 mm Plain-end Culture Tubes
38 mm Plain-end Culture Tubes
Unit Size:
Pack of 1000
Pack of 250
Pack of 500
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color For Use With (Equipment) Unit Size
10399924 Natural 16 mm Plain-end Culture Tubes Pack of 1000
10799335 Natural 20 mm Plain-end Culture Tubes Pack of 500
11758283 Natural 13 mm Plain-end Culture Tubes Pack of 500
10349594 Natural 38 mm Plain-end Culture Tubes Pack of 250
10389354 Red 13 mm Plain-end Culture Tubes Pack of 1000
10616285 Green 13 mm Plain-end Culture Tubes Pack of 1000
10647245 Yellow 13 mm Plain-end Culture Tubes Pack of 1000
10084554 Blue 13 mm Plain-end Culture Tubes Pack of 1000
10024174 Red 16 mm Plain-end Culture Tubes Pack of 1000
10369934 Yellow 16 mm Plain-end Culture Tubes Pack of 1000
10451815 Green 16 mm Plain-end Culture Tubes Pack of 1000
10626285 Green 18 mm Plain-end Culture Tubes Pack of 1000
10492375 Blue 18 mm Plain-end Culture Tubes Pack of 1000
10257774 Yellow 18 mm Plain-end Culture Tubes Pack of 1000
10638205 Yellow 25 mm Plain-end Culture Tubes Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Product Code. 10399924 Supplier DWK Life Sciences Supplier No. 7366016
  • ONLINE EXCLUSIVE PRICE
    139.70 EUR
    124.92 EUR / Pack of 1000
    Save 14.78 EUR
    10% Off
Estimated Shipment: 19-04-2027
Available in natural (clear) or choice of four colors
  • For plain-end culture tubes
  • Keep tube contents sterile without need for cotton plugging
  • Polypropylene
  • Autoclavable
Specifications
Product Type Push Cap
Autoclavable Autoclavable
Material Polypropylene
Color Natural
Sterility Non-sterile
For Use With (Equipment) 16 mm Plain-end Culture Tubes
Disposable Reusable