missing translation for 'onlineSavingsMsg'
Learn More

Kifunensine, >98%, MP Biomedicals™

Product Code. 11460132 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460132 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11460132 Supplier MP Biomedicals Supplier No. 0215995201
  • ONLINE EXCLUSIVE PRICE
    244.00 EUR
    212.10 EUR / 1mg
    Save 31.90 EUR
    13% Off
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 109944-15-2
Color White
Assay Percent Range >98%
Molecular Formula C8H12N2O6
Quantity 1 mg
Synonym kifunensine, +-kifunensine, kifunensine, kitasatosporia kifunense, 5r,6r,7s,8r,8as-6,7,8-trihydroxy-5-hydroxymethyl hexahydroimidazo 1,2-a pyridine-2,3-dione, kif, 5r,6r,7s,8r,8as-6,7,8-trihydroxy-5-hydroxymethyl-1,5,6,7,8,8a-hexahydroimidazo 1,2-a pyridine-2,3-dione, 5r,6r,7s,8r,8as-hexahydro-6,7,8-trihydroxy-5-hydroxymethyl-imidazo 1,2-a pyridine-2,3-dione, imidazo 1,2-a pyridine-2,3-dione, hexahydro-6,7,8-trihydroxy-5-hydroxymethyl-, 5r-5alpha,6beta,7alpha,8alpha,8aalpha, 2wvz, d0ad9y
InChI Key OIURYJWYVIAOCW-PQMKYFCFSA-N
SMILES C(C1C(C(C(C2N1C(=O)C(=O)N2)O)O)O)O
IUPAC Name (5R,6R,7S,8R,8aS)-6,7,8-trihydroxy-5-(hydroxymethyl)-1,5,6,7,8,8a-hexahydroimidazo[1,2-a]pyridine-2,3-dione
Molecular Weight (g/mol) 232.192
PubChem CID 130611
Formula Weight 232.2
Physical Form Powder
Chemical Name or Material Kifunensine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.