missing translation for 'onlineSavingsMsg'
Learn More

Kevetrin hydrochloride, MedChemExpress

Product Code. p-200098060
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16467076 100 mg 100mg
16463067 50 mg 50mg
16457018 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16467076 Supplier Medchem Express Supplier No. HY16271

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Kevetrin hydrochloride is a small molecule and activator of the tumor suppressor protein p53, with potential antineoplastic activity.

Kevetrin hydrochloride is a small molecule and activator of the tumor suppressor protein p53, with potential antineoplastic activity.

Specifications

Chemical Name or Material Kevetrin hydrochloride
Physical Form Solid
CAS 66592-89-0
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C5H10ClN3S
For Use With (Application) Cancer-programmed cell death
Synonym 4-Isothioureidobutyronitrile hydrochloride thioureidobutyronitrile hydrochloride thioureido butyronitrile hydrochloride
Solubility Information H2O : 100 mg/mL (556.58 mM; Need ultrasonic) ∣DMSO : ≥ 40 mg/mL (222.63 mM)
SMILES NC(SCCCC#N)=N.[H]Cl
Molecular Weight (g/mol) 179.67
Formula Weight 179.67
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.