missing translation for 'onlineSavingsMsg'
Learn More

Ketoprofen , MP Biomedicals

Product Code. 11403352 Shop All MP Biomedicals Products
Change view
Click to view available options
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11403352 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11403352 Supplier MP Biomedicals Supplier No. 0215515480

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifica

CAS 22071-15-4
Color White
Molecular Formula C16H14O3
MDL Number MFCD00055790
Quantity 100 g
Synonym ketoprofen, 2-3-benzoylphenyl propanoic acid, orudis, m-benzoylhydratropic acid, 2-3-benzoylphenyl propionic acid, ketoprofene, profenid, oruvail, 3-benzoylhydratropic acid, alrheumat
InChI Key DKYWVDODHFEZIM-UHFFFAOYSA-N
SMILES CC(C1=CC=CC(=C1)C(=O)C2=CC=CC=C2)C(=O)O
IUPAC Name 2-(3-benzoylphenyl)propanoic acid
Molecular Weight (g/mol) 254.285
PubChem CID 3825
ChEBI CHEBI:6128
Formula Weight 254.28
Physical Form Powder
Chemical Name or Material Ketoprofen, Molecular biology
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.