Learn More
Hamilton™ Kel-F Hub Needle
Product Code. 11572445
Change view
Click to view available options
Gauge:
22 G
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Gauge Unit Size
11572445 22 G Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11572445 Supplier Hamilton™ Supplier No. 775013
  • ONLINE EXCLUSIVE PRICE
    215.00 EUR
    191.45 EUR / Pack of 6
    Save 23.55 EUR
    10% Off
Estimated Shipment: 24-11-2026
Specifications
Product Type Hub Needle
Gauge 22 G
Hub Material Kel-F
Length (Metric) 9.5 to 304.8 mm
Style Custom Point