missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ KC7F2

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Inhibitor of HIF-1α. Thought to act via down-regulation of HIF-1α protein synthesis; reduces phosphorylation of eIF4E binding protein 1 (4EBP1) and p70 S6K in hypoxic conditions. Also blocks hypoxia-induced HIF-1α accumulation in a range of human cancer cell lines. Inhibits the expression of HIF target genes, such as carbonic anhydrase IX and MMP2.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 570.38 |
| Purity | >98% |
| Molecular Weight (g/mol) | 570.354 |
| Chemical Name or Material | KC7F2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction