missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KB-R7943 mesylate, MedChemExpress
Description
KB-R7943 mesylate is a widely used inhibitor of the reverse Na+/Ca2+ exchanger (NCXrev) with IC50 of 5.7±2.1 μM. KB-R7943 mesylate induces cancer cell death via activating the JNK pathway and blocking autophagic flux.
Specifications
Specifications
| Chemical Name or Material | KB-R7943 mesylate |
| CAS | 182004-65-5 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C17H21N3O6S2 |
| Quantity | 10 mg |
| Solubility Information | DMSO : ≥ 27 mg/mL (63.16 mM) ∣H2O : 4.3 mg/mL (10.06 mM; Need warming) |
| SMILES | NC(SCCC1=CC=C(OCC2=CC=C([N+]([O-])=O)C=C2)C=C1)=N.CS(=O)(O)=O |
| Molecular Weight (g/mol) | 427.5 |
| Formula Weight | 427.5 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?