missing translation for 'onlineSavingsMsg'
Learn More

kb NB 142-70, MedChemExpress

Product Code. 16476598
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16476598

Brand: Medchem Express HY15528

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress kb NB 142-70 is a potent PKD inhibitor, with IC50s of 28.3, 58.7 and 53.2 nM for PKD1, PKD2, and PKD3, respectively. kb NB 142-70 also has antitumor activity.

kb NB 142-70 is a potent PKD inhibitor, with IC50s of 28.3, 58.7 and 53.2 nM for PKD1, PKD2, and PKD3, respectively. kb NB 142-70 also has antitumor activity.

Specifications

Chemical Name or Material kb NB 142-70
CAS 1233533-04-4
Color Light Gray
Purity Grade Notes Research
Molecular Formula C11H9NO2S2
Quantity 10 mg
Solubility Information DMSO : 46.67 mg/mL (185.70 mM; Need ultrasonic)
SMILES O=C1NCCSC2=C1SC3=CC=C(O)C=C32
Molecular Weight (g/mol) 251.32
Formula Weight 251.32
Percent Purity 98.85%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.