Learn More
Nordvlies Katrin System Electric Towel Dispenser
Product Code. 15741097
Change view
Click to view available options
Color:
White
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15741097 White Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15741097 Supplier Nordvlies Supplier No. 93701
  • ONLINE EXCLUSIVE PRICE
    327.00 EUR
    261.46 EUR / Pack of 1
    Save 65.54 EUR
    20% Off
Estimated Shipment: 09-04-2027
System Electric Towel Dispenser is fast and easy to use.
  • A full range of Katrin hand drying roll tissue qualities available.
  • Fast and easy to use; sheet always available.
  • High capacity
    • 800 sheets to a roll is enough to dry hands 400 to 530 times
    • Ideal in high traffic areas.
  • Feeds the next sheet automatically.
  • Designed to ensure all the paper on the roll is used.
  • Very low power consumption
    • Powered by 4 x LR20 1.5 V batteries (not supplied).
  • Controlled consumption for high traffic washrooms.
  • Main raw materials of major plastic parts: ABS (Acrylonitrile butadiene styrene), PC (polycarbonate)
Specifications
Product Type Towel Dispenser
Color White
Material ABS, Polycarbonate