Learn More
Thermo Scientific™ KaryoStat+™ Genetic Stability Assay Service
Product Code. 17882606
Change view
Click to view available options
1 Sample
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17882606 1 Sample
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17882606 Supplier Thermo Scientific™ Supplier No. A52849
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
KaryoStat+ Genetic Stability Assay Service is a fast and cost-effective alternative to typical G-banded karyotyping.
  • A fast and cost-effective alternative to typical G-banded karyotyping
  • Leverages the array-based CytoScan HT-CMA 96F Assay Kit (906025), offering whole-genome coverage for accurate detection of chromosomal aberrations in human primary and stem cells
The KaryoStat+ Assay Service offers:
  • Fast data delivery – turnaround time of three weeks or less
  • Convenient sample prep – our assay uses frozen cell pellet, eliminating need for shipping live cells
  • Higher resolution – microarray resolution five times higher than traditional G-banding
  • Exceptional support – a dedicated project manager for frequent project communication
  • Broad cell type compatibility – all non-transformed human cell types accepted
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 sample
Certifications/Compliance ISO 9001
Duration 3 Weeks
Product Type Genetic Stability Assay Service
Product Line Applied Biosystems
Field Research Use Only
Target Audience Pluripotent Stem Cell Researchers
Species Human