Learn More
Thermo Scientific™ KaryoStat+™ with Cell ID Assay Service
Product Code. 17892606 Shop All Applied Biosystems Products
Change view
Click to view available options
1 Sample
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17892606 1 Sample
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17892606 Supplier Thermo Scientific™ Supplier No. A52986
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
KaryoStat+ with Cell ID Assay Service is a fast and cost-effective alternative to typical G-banded karyotyping that evaluates both genetic stability and SNP-based cell identification.
  • A fast and cost-effective alternative to typical G-banded karyotyping that evaluates both genetic stability and SNP-based cell identification
  • KaryoStat+ service leverages the array-based CytoScan HT-CMA 96F Assay Kit (906025), offering whole-genome coverage for accurate detection of chromosomal aberrations in human primary and stem cells
  • Cell ID leverages the KaryoStat Assay (905403) to compare genotypes across samples
The KaryoStat+ with Cell ID Assay Service offers:
  • Fast data delivery – turnaround time of four weeks or less
  • Convenient sample prep – assay uses frozen cell pellet, eliminating need for shipping live cells
  • Higher resolution – microarray resolution five times higher than traditional G-banding
  • Exceptional support – a dedicated project manager for frequent project communication
  • Broad cell type compatibility – all non-transformed human cell types accepted
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 sample
Duration 3 Weeks
Product Type Assay Service
Product Line Applied Biosystems
Field Research Use Only
Target Audience Stem Cell and Immuno-oncology Researchers
Species Human