missing translation for 'onlineSavingsMsg'
Learn More

Karetazan, TRC

Product Code. 30135959
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135959 50 mg 50mg
30138418 25 mg 25mg
30011239 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135959 Supplier Toronto Research Chemicals Supplier No. TRCK45225550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81051-65-2
Molecular Formula C15 H14 Cl N O3
Molecular Weight (g/mol) 291.73
Synonym 2-(4-Chlorophenyl)-1-ethyl-1,4-dihydro-6-methyl-4-oxo-3-pyridinecarboxylic acid, Karetazan
IUPAC Name 2-(4-chlorophenyl)-1-ethyl-6-methyl-4-oxopyridine-3-carboxylic acid
SMILES CCN1C(=CC(=O)C(=C1c2ccc(Cl)cc2)C(=O)O)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C15H14ClNO3/c1-3-17-9(2)8-12(18)13(15(19)20)14(17)10-4-6-11(16)7-5-10/h4-8H,3H2,1-2H3,(H,19,20)
Formula Weight 291.0662
Chemical Name or Material Karetazan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.