Learn More
Karetazan 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30323402
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30323402 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30323402 Supplier Dr. Ehrenstorfer Supplier No. DREA14511000AL100
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Karetazan
CAS 81051-65-2
Analyte or Component Names Karetazan
Shipping Condition +20°C
Molecular Formula C15 H14 Cl N O3
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym 2-(4-Chlorophenyl)-1-ethyl-1,4-dihydro-6-methyl-4-oxo-3-pyridinecarboxylic Acid
InChI Formula InChI=1S/C15H14ClNO3/c1-3-17-9(2)8-12(18)13(15(19)20)14(17)10-4-6-11(16)7-5-10/h4-8H,3H2,1-2H3,(H,19,20)
SMILES CCN1C(=CC(=O)C(=C1c2ccc(Cl)cc2)C(=O)O)C
IUPAC Name 2-(4-chlorophenyl)-1-ethyl-6-methyl-4-oxopyridine-3-carboxylic acid
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 291.73
Quantity 1 mL
Formula Weight 291.0662 g/mol